Learn More
Agilent BioTek Joystick Controller for Imaging Multi-Mode Reader
Product Code. 15468722 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Cytation 5 and Lionheart Imaging Readers
Unit Size:
Each
Product Code. 15468722
Brand: Agilent BioTek 1320003
For use with Cytation 5 Cell Imaging Multi-Mode Readers.
For use with Cytation 5 Cell Imaging Multi-Mode Readers.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Joystick Controller
For Use With (Equipment) Cytation 5 and Lionheart Imaging Readers