missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JIP2 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £359.00
Specifications
| Antigen | JIP2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18314254
|
Novus Biologicals
NBP3-09334-25UL |
25 μg |
£161.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18360677
|
Novus Biologicals
NBP3-09334-100UL |
100 μg |
£359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JIP2 Polyclonal specifically detects JIP2 in Rat samples. It is validated for Western Blot.Specifications
| JIP2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS buffer, 2% sucrose | |
| 23542 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| C-Jun-amino-terminal kinase-interacting protein 2, homologous to mouse JIP-1, IB-2, IB2JIP2Mitogen-activated protein kinase 8-interacting protein 2, islet-brain 2, Islet-brain-2, JIP-2, JNK MAP kinase scaffold protein 2, JNK MAP kinase scaffold protein JIP2, JNK-interacting protein 2, mitogen-activated protein kinase 8 interacting protein 2, PRKM8 interacting protein-like, PRKM8IPL | |
| The immunogen is a synthetic peptide directed towards the middle region of Rat JIP2 (XP_001055248). Peptide sequence SEPEPEPEPEPLHEPPRRPAFLPVGQDDTNSEYESGSESEPDLSEDADSP | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title