missing translation for 'onlineSavingsMsg'
Learn More

Jenway™ Bench Meters

Product Code. 12706658
Change view
Click to view available options
Outputs:
Analogue and RS232
Analogue, RS232 and IrDA
Power Supply:
UK
Probe Type:
ATC
Conductivity
DO2
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Outputs Power Supply Probe Type unitSize
12706658 Analogue and RS232 UK ATC Each
12983237 Analogue and RS232 UK ATC Each
12993237 Analogue, RS232 and IrDA UK ATC Each
12756965 Analogue and RS232 UK Conductivity Each
12766965 Analogue, RS232 and IrDA UK Conductivity Each
13141794 Analogue, RS232 and IrDA UK DO2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12706658 Supplier Jenway™ Supplier No. 3545901
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use this versatile benchtop meter for a variety of applications. Jenway™ Bench Meters are ideal for routine analysis.

The 3510 is a versatile, simple to use pH, mV and temperature meter that is ideal for routine analysis. With up to a three decimal place resolution and a choice of up to three calibration points, the 3510 provides the user with added flexibility where demands for enhanced performance may be required. A choice of pH calibration buffers to DIN, JIS and NIST standards can be used for automatic calibration, as well as manually entered buffer values. The pH measurement meter also has the ability to store up to 32 results.
  • Simultaneous readout of pH and temperature
  • pH resolution to three decimal places
  • One, two or three point calibration
  • Automatic or manual buffer selection
  • DIN, JIS and NIST buffer standards can be used for automatic calibration
  • Storage of up to 32 results
  • RS232 connection to printer or PC via DataWay

The 3520 is a dynamic pH/mV meter making it suitable for the broadest range of research, general laboratory, QC and GLP based applications. The 3520 pH/mV meter offers research grade specifications with a comprehensive range of features and functions. The built-in data logger can store, print or output readings based on a wide range of trigger options, making this model suitable for monitoring and controlling pH levels.
  • Ideal for quality control and GLP applications
  • Up to three decimal place resolution
  • 1, 2 or 3 point pH calibration
  • Automatic or manual buffer selection
  • Multiple language options
  • Storage of up to 500 results
  • RS232 connection to printer or PC or infra red (IrDA) communication

The 4510 is an easy to use bench conductivity meter with the flexibility to meet the broadest range of user requirements. For those applications where greater accuracy is required the 4510 has automatic conductivity standard recognition and endpoint detection. Set up options include cell constant, temperature coefficient and reference temperature. The instrument can store up to 32 readings, which can also be sent to a printer or transferred to computer via DataWay and the RS232 interface.
  • Simultaneous display of conductivity or TDS and temperature
  • Automatic selection of range for best resolution
  • Calibration by cell constant or standard solutions
  • Automatic standard recognition
  • Storage of up to 32 results
  • Bi-directional RS232 link to printer or PC

The 4520 is a high specification conductivity meter with a host of measurement modes. The 4520 laboratory meter offers modes for conductivity/resistivity/TDS/salinity and temperature and provides additional accuracy with 1, 2 or 3 point conductivity calibration across the wide measurement range of 0 to 19.99S. The dedicated 'pure water' mode ensures the optimum accuracy for this difficult application.
  • Conductivity, resistivity, TDS and salinity modes
  • Special pure water mode for ultra low conductivity measurements
  • Storage of 500 results with built-in datalogger
  • Infrared data link (IrDA) for connection to printer and RS232 link to connect to printer or PC via DataWay software

Specifications

Outputs Analogue and RS232
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.003
Calibration Points 1 to 3
Display Type Dual
Range (pH) -2.000 to 19.998
Resolution (pH) 0.001/0.01/0.0
Temperature Range (Metric) 0°C to 100°C (ATC/Manual)
For Use With (Application) routine analysis
Includes Electrode stand and holder, ATC probe, BNC shorting plug, pH 4, 7, and 10 buffer sachets and UK power supply
Log Function Type 32 readings
Model 3510
Power Supply UK
Probe Type ATC
Product Type pH/mV/Temperature
Range (Relative mV) –1.999.9 to 1999.9mV
Resolution (mV) 0.1 to 1mV
Temperature Accuracy ±0.5°C
Temperature Resolution 0.1°C
Length (Metric) 210 mm
Width (Metric) 210 mm
Height (Metric) 55 mm
Electrical Requirements 9VAC ±10% at 50/60Hz
Voltage 9 V
Frequency 50/60Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.