missing translation for 'onlineSavingsMsg'
Learn More

(±)-Jasmonic Acid, TRC

Product Code. 17991725
Change view
Click to view available options
Quantity:
25 mg
50 mg
75 mg
Unit Size:
25mg
50mg
75mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991725 25 mg 25mg
17962385 75 mg 75mg
17923535 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17991725 Supplier Toronto Research Chemicals Supplier No. TRCJ21051025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3572-66-5
Molecular Formula C12 H18 O3
Molecular Weight (g/mol) 210.27
Synonym 3-Oxo-2-((Z)-2-pentenyl)cyclopentane-1-acetic Acid, 3-Oxo-2-(2-penten-1-yl)cyclopentaneacetic Acid, 3-Oxo-2-(2-pentenyl)-cyclopentaneacetic Acid, 3-Oxo-2-(2-penten-1-yl)-cyclopentaneacetic Acid
IUPAC Name 2-[3-oxo-2-[(E)-pent-2-enyl]cyclopentyl]acetic acid
SMILES CC\C=C\CC1C(CC(=O)O)CCC1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H18O3/c1-2-3-4-5-10-9(8-12(14)15)6-7-11(10)13/h3-4,9-10H,2,5-8H2,1H3,(H,14,15)/b4-3+
Formula Weight 210.1256
Percent Purity >95%
Chemical Name or Material (±)-Jasmonic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.