CAS RN 3572-66-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(±)-Jasmonic Acid, TRC
CAS: 3572-66-5 Molecular Formula: C12 H18 O3 Molecular Weight (g/mol): 210.27 Synonym: 3-Oxo-2-((Z)-2-pentenyl)cyclopentane-1-acetic Acid,3-Oxo-2-(2-penten-1-yl)cyclopentaneacetic Acid,3-Oxo-2-(2-pentenyl)-cyclopentaneacetic Acid,3-Oxo-2-(2-penten-1-yl)-cyclopentaneacetic Acid IUPAC Name: 2-[3-oxo-2-[(E)-pent-2-enyl]cyclopentyl]acetic acid SMILES: CC\C=C\CC1C(CC(=O)O)CCC1=O