missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Jarid1A Antibody (SR3825), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | Jarid1A |
|---|---|
| Clone | SR3825 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1:80 |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041776
|
Novus Biologicals™
NBP3-27810-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041690
|
Novus Biologicals™
NBP3-27810-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Jarid1A Monoclonal antibody specifically detects Jarid1A in Human,Mouse samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| Jarid1A | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1:80 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1A, JARID1AJumonji, AT rich interactive domain 1A (RBP2-like), Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji/ARID domain-containing protein 1A, KDM5A, lysine (K)-specific demethylase 5A, RBBP-2, RBBP2lysine-specific demethylase 5A, RBP2retinoblastoma binding protein 2, Retinoblastoma-binding protein 2jumonji, AT rich interactive domain 1A | |
| A synthesized peptide derived from human Jarid1A (Uniprot #: P29375) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3825 | |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 5927 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title