missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Jarid1A Antibody (SR3825), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27810-50ul
This item is not returnable.
View return policy
Description
Jarid1A Monoclonal antibody specifically detects Jarid1A in Human,Mouse samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Jarid1A | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR3825 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1:80 | |
| EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1A, JARID1AJumonji, AT rich interactive domain 1A (RBP2-like), Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji/ARID domain-containing protein 1A, KDM5A, lysine (K)-specific demethylase 5A, RBBP-2, RBBP2lysine-specific demethylase 5A, RBP2retinoblastoma binding protein 2, Retinoblastoma-binding protein 2jumonji, AT rich interactive domain 1A | |
| A synthesized peptide derived from human Jarid1A (Uniprot #: P29375) | |
| 50 μL | |
| Cell Biology | |
| 5927 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction