missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jagged 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £360.00
Specifications
| Antigen | Jagged 2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 5-10ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18108353
|
Novus Biologicals
NBP2-27420SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18105942
|
Novus Biologicals
NBP2-27420 |
0.1 mg |
£360.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Jagged 2 Polyclonal specifically detects Jagged 2 in Human, Primate samples. It is validated for Western Blot.Specifications
| Jagged 2 | |
| Western Blot 5-10ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| Q9Y219 | |
| 3714 | |
| A portion of amino acids 150-210 of human JAG2 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Stem Cell Signaling Pathway | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| HJ2, jagged 2, Jagged2, protein jagged-2, SER2 | |
| JAG2 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human and monkey, 94% homologous in mouse, rat and horse, 76% homologous in opossum and 70% homologous in chicken. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title