missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jagged 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27420SS
This item is not returnable.
View return policy
Description
Jagged 2 Polyclonal specifically detects Jagged 2 in Human, Primate samples. It is validated for Western Blot.
Specifications
| Jagged 2 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| HJ2, jagged 2, Jagged2, protein jagged-2, SER2 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and monkey, 94% homologous in mouse, rat and horse, 76% homologous in opossum and 70% homologous in chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 5-10ug/ml | |
| Q9Y219 | |
| JAG2 | |
| A portion of amino acids 150-210 of human JAG2 was used as the immunogen. | |
| 0.025 mg | |
| Cell Cycle and Replication, Stem Cell Signaling Pathway | |
| 3714 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction