missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ JAG1 (Human) Recombinant Protein (Q01)

Human JAG1 partial ORF with GST-tag at N-terminal

Brand:  Abnova™ H00000182-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16013605

  • £425.00 / 25µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.

  • Theoretical MW: 35.64kDa
  • Preparation Method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Best use within three months from the date of receipt of this protein

Enzyme Linked Immunosorbent Assay, Western Blotting, Antibody Production, Protein Array

Specifications

Specifications

NP_000205
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
35.64
8
Glutathione Sepharose 4 Fast Flow
25 ug
PNPCQNGAQCYNRASDYFCKCPEDYEGKNCSHLKDHCRTTPCEVIDSCTVAMASNDTPEGVRYISSNVCGPHGKCKSQSGGKFTCDCNKG
AGS/AHD/AWS/CD339/HJ1/JAGL1/MGC104644
JAG1
Wheat Germ (in vitro)
GST
Antibody Production, ELISA, Protein Array, Western Blot
182
JAG1 (Human) Recombinant Protein (Q01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
JAG1
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers