missing translation for 'onlineSavingsMsg'
Learn More

Jaceosidin, TRC

Product Code. 30135287
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135287 100 mg 100mg
30134258 50 mg 50mg
17967544 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135287 Supplier Toronto Research Chemicals Supplier No. TRCJ125450100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18085-97-7
Molecular Formula C17H14O7
Molecular Weight (g/mol) 330.29
Synonym 4',5,7-Trihydroxy-3',6-dimethoxy-flavone, 5,7-Dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 3',6-Dimethoxy-4',5,7-trihydroxyflavone, 4',5,7-Trihydroxy-3',6-dimethoxyflavone, 4'-Demethyleupatilin, 5,7,4'-Trihydroxy-6,3'-dimethoxyflavone;
IUPAC Name 5,7-dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-6-methoxychromen-4-one
SMILES COc1cc(ccc1O)C2=CC(=O)c3c(O)c(OC)c(O)cc3O2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H14O7/c1-22-13-5-8(3-4-9(13)18)12-6-10(19)15-14(24-12)7-11(20)17(23-2)16(15)21/h3-7,18,20-21H,1-2H3
Formula Weight 330.074
Percent Purity >95%
Chemical Name or Material Jaceosidin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.