Learn More
Ivermectin, Thermo Scientific™
£60.65
Chemical Identifiers
| CAS | 70288-86-7 |
|---|---|
| Molecular Formula | C48H74O14 |
| Molecular Weight (g/mol) | 875.106 |
| MDL Number | MFCD00869511 |
| InChI Key | AZSNMRSAGSSBNP-RDKIRRGNSA-N |
| Synonym | ivermectin |
| PubChem CID | 122172888 |
| SMILES | CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15435079
|
Thermo Scientific Alfa Aesar
J62777.03 |
1g |
£67.50 £60.65 / 1g Save £6.85 10% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as a positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor.
Solubility
Soluble in methyl ethyl ketone (50 mg/ml), and DMSO (50 mM).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
Chemical Identifiers
| 70288-86-7 | |
| 875.106 | |
| AZSNMRSAGSSBNP-RDKIRRGNSA-N | |
| 122172888 |
| C48H74O14 | |
| MFCD00869511 | |
| ivermectin | |
| CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C |
Specifications
| 70288-86-7 | |
| MFCD00869511 | |
| 14,5248 | |
| Soluble in methyl ethyl ketone (50mg/ml),and DMSO (50 mM). | |
| CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C | |
| 122172888 | |
| Ivermectin |
| C48H74O14 | |
| UN2811 | |
| ivermectin | |
| AZSNMRSAGSSBNP-RDKIRRGNSA-N | |
| 875.106 | |
| 875.11 |
Safety and Handling
GHS H Statement
H300-H360-H319
Fatal if swallowed.
May damage fertility or the unborn child.
Causes serious eye irritation.
P201-P202-P264b-P270-P280i-P281-P301+P310-P305+P351+P338-P308+P313-P330-P501c
H300-H319-H360
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 274-536-0
missing translation for 'rtecsNumber' : IH7891500
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.