missing translation for 'onlineSavingsMsg'
Learn More

Ivermectin

Product Code. 15435079
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435079 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15435079 Supplier Thermo Scientific Alfa Aesar Supplier No. J62777.03
  • ONLINE EXCLUSIVE PRICE
    £71.60
    £60.65 / 1g
    Save £10.95
    15% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 70288-86-7 | C48H74O14 | 875.106 g/mol

It is used as a positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor.

Solubility
Soluble in methyl ethyl ketone (50 mg/ml), and DMSO (50 mM).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70288-86-7
Molecular Formula C48H74O14
Molecular Weight (g/mol) 875.106
MDL Number MFCD00869511
InChI Key AZSNMRSAGSSBNP-RDKIRRGNSA-N
Synonym ivermectin
PubChem CID 122172888
SMILES CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C

Specifications

Color White
Quantity 1 g
UN Number UN2811
Merck Index 14,5248
Solubility Information Soluble in methyl ethyl ketone (50mg/ml),and DMSO (50 mM).
Formula Weight 875.11
Physical Form Powder
Chemical Name or Material Ivermectin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.