missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITPK1 Recombinant Rabbit Monoclonal Antibody (23GB5995), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | ITPK1 |
|---|---|
| Clone | 23GB5995 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089508
![]() |
Invitrogen™
MA552966 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089494
|
Invitrogen™
MA552967 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITPK1 Recombinant Monoclonal antibody specifically detects ITPK1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ITPK1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13572 | |
| 3705 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5995 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BC031182; inositol 1,3,4-triphosphate 5/6 kinase; inositol 1,3,4-trisphosphate 5/6-kinase; Inositol-tetrakisphosphate 1-kinase; inositol-triphosphate 5/6-kinase; ins(1,3,4)P(3) 5/6-kinase; ITPK1; ITRPK1 | |
| A synthesized peptide derived from human ITPK1 (362-402AA). | |
| Primary | |
| -20°C | |
| ITPK1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title