missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITPK1 Recombinant Rabbit Monoclonal Antibody (23GB5995), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552966
This item is not returnable.
View return policy
Description
ITPK1 encodes an enzyme that belongs to the inositol 1,3,4-trisphosphate 5/6-kinase family. This enzyme regulates the synthesis of inositol tetraphosphate, and downstream products, inositol pentakisphosphate and inositol hexakisphosphate. Inositol metabolism plays a role in the development of the neural tube. Disruptions in this gene are thought to be associated with neural tube defects. A pseudogene of this gene has been identified on chromosome X.
Specifications
| ITPK1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ITPK1 | |
| BC031182; inositol 1,3,4-triphosphate 5/6 kinase; inositol 1,3,4-trisphosphate 5/6-kinase; Inositol-tetrakisphosphate 1-kinase; inositol-triphosphate 5/6-kinase; ins(1,3,4)P(3) 5/6-kinase; ITPK1; ITRPK1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3705 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5995 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13572 | |
| ITPK1 | |
| A synthesized peptide derived from human ITPK1 (362-402AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido