missing translation for 'onlineSavingsMsg'
Learn More

ITIH3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£178.00 - £255.00

Specifications

Antigen ITIH3
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18680642
View Documents
Novus Biologicals
NBP2-97130-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18698901
View Documents
Novus Biologicals
NBP2-97130-50ul
50 μg
£178.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ITIH3 Polyclonal antibody specifically detects ITIH3 in Human samples. It is validated for ELISA
Specifications

Specifications

ITIH3
ELISA
Unconjugated
Rabbit
Human
H3 polypeptide, inter-alpha (globulin) inhibitor H3, inter-alpha (globulin) inhibitor, H3 polypeptide, inter-alpha-inhibitor heavy chain 3, inter-alpha-trypsin inhibitor heavy chain H3, ITI heavy chain H3, ITI-HC3, serum-derived hyaluronan-associated protein, SHAP
Produced in rabbits immunized with purified, recombinant Human ITIH3 (Accession#: NP_002208.3; Met1-Asp651)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
0.2 um filtered solution in PBS
3699
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.