missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITIH3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97130-100ul
This item is not returnable.
View return policy
Description
ITIH3 Polyclonal antibody specifically detects ITIH3 in Human samples. It is validated for ELISA
Specifications
| ITIH3 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| H3 polypeptide, inter-alpha (globulin) inhibitor H3, inter-alpha (globulin) inhibitor, H3 polypeptide, inter-alpha-inhibitor heavy chain 3, inter-alpha-trypsin inhibitor heavy chain H3, ITI heavy chain H3, ITI-HC3, serum-derived hyaluronan-associated protein, SHAP | |
| Produced in rabbits immunized with purified, recombinant Human ITIH3 (Accession#: NP_002208.3; Met1-Asp651) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 3699 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction