missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGB6 Monoclonal Antibody (C5), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543682
This item is not returnable.
View return policy
Description
ITGB6 (Integrin alpha-V/beta-6) is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalization of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion.
Specifications
| ITGB6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P18564 | |
| Itgb6 | |
| Tyr131-Leu371 (Accession P18564), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C5 | |
| Unconjugated | |
| Itgb6 | |
| 2210409C20Rik; 4831415H04Rik; AI1H; integrin beta 6; integrin beta-6; integrin subunit beta 6; integrin, beta 6; Itgb6 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3694 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction