missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGB6 Monoclonal Antibody (C5), Invitrogen™

Mouse Monoclonal Antibody
£178.00 - £387.00
Specifications
| Antigen | ITGB6 |
|---|---|
| Clone | C5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17674136
|
Invitrogen™
MA543683 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17664136
![]() |
Invitrogen™
MA543682 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITGB6 (Integrin alpha-V/beta-6) is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalization of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion.Specifications
| ITGB6 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 2210409C20Rik; 4831415H04Rik; AI1H; integrin beta 6; integrin beta-6; integrin subunit beta 6; integrin, beta 6; Itgb6 | |
| Itgb6 | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| C5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P18564 | |
| 3694 | |
| Tyr131-Leu371 (Accession P18564), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Itgb6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title