missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITCH/AIP4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55083-25ul
This item is not returnable.
View return policy
Description
ITCH/AIP4 Polyclonal specifically detects ITCH/AIP4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ITCH/AIP4 | |
| Polyclonal | |
| Western Blot 1:250 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ITCH | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GFTGASQNDDGSRSKDETRVSTNGSDDPEDAGAGENRRVSGNNSPSLSNGGFKPSRPPRPSRPPPPTPRRPASVNGS | |
| 25 μL | |
| Cell Cycle and Replication, Chromatin Research, DNA Repair | |
| 83737 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction