missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISYNA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54984
This item is not returnable.
View return policy
Description
ISYNA1 Polyclonal specifically detects ISYNA1 in Human samples. It is validated for Western Blot.
Specifications
| ISYNA1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 5.5.1.4, hINO1, hIPS, INO1, INOS, inositol-3-phosphate synthase 1, IPSIPS 1, MI-1-P synthase, MIP synthase, Myo-inositol 1-phosphate synthase A1, Myo-inositol-1-phosphate synthase | |
| Rabbit | |
| 61 kDa | |
| 100 μL | |
| Apoptosis | |
| 51477 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NPH2 | |
| ISYNA1 | |
| Synthetic peptides corresponding to ISYNA1(myo-inositol 1-phosphate synthase A1) The peptide sequence was selected from the N terminal of ISYNA1. Peptide sequence LQEQLWPHMEALRPRPSVYIPEFIAANQSARADNLIPGSRAQQLEQIRRD. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat: 86%; Equine: 86%; Mouse: 86%; Bovine: 86%. | |
| Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Yeast | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction