missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isradipine, 98+%, Thermo Scientific™
L-type calcium channel blocker
£100.00 - £333.99
Chemical Identifiers
| CAS | 75695-93-1 |
|---|---|
| Molecular Formula | C19H21N3O5 |
| Molecular Weight (g/mol) | 371.39 |
| MDL Number | MFCD00153820 |
| InChI Key | HMJIYCCIJYRONP-UHFFFAOYNA-N |
| Synonym | isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine |
| PubChem CID | 3784 |
| IUPAC Name | 3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| SMILES | COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15426079
|
Thermo Scientific Alfa Aesar
J63920.MA |
10mg |
£102.00 £100.00 / 10mg Save £2.00 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15436079
|
Thermo Scientific Alfa Aesar
J63920.MF |
50mg |
£341.00 £333.99 / 50mg Save £7.01 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIsradipine is a L-type calcium channel blocker, Isradipine has anti-atherosclerotic effects in animals and improves endothelium-mediated nitric oxide (NO)-dependent vasodilation in vitro. Additional studies indicate that Isradipine may provide a neuroprotective effect in a rat model of focal ischemia and that Isradipine produces a dose-dependent sparing of dopamine neuronal fibers and cell bodies.
Solubility
Soluble in DMSO
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
| 75695-93-1 | |
| 371.39 | |
| HMJIYCCIJYRONP-UHFFFAOYNA-N | |
| 3784 | |
| COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C |
| C19H21N3O5 | |
| MFCD00153820 | |
| isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine | |
| 3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
Specifications
| 75695-93-1 | |
| MFCD00153820 | |
| isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine | |
| HMJIYCCIJYRONP-UHFFFAOYNA-N | |
| 3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate | |
| 3784 | |
| ≥98% |
| C19H21N3O5 | |
| 14,5240 | |
| Soluble in DMSO | |
| COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C | |
| 371.39 | |
| 371.39 | |
| Isradipine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title