missing translation for 'onlineSavingsMsg'
Learn More

Isradipine, 98+%

Product Code. 15436079
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436079 50 mg 50mg
15426079 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436079 Supplier Thermo Scientific Alfa Aesar Supplier No. J63920.MF
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-type calcium channel blocker | CAS: 75695-93-1 | C19H21N3O5 | 371.39 g/mol

Isradipine is a L-type calcium channel blocker, Isradipine has anti-atherosclerotic effects in animals and improves endothelium-mediated nitric oxide (NO)-dependent vasodilation in vitro. Additional studies indicate that Isradipine may provide a neuroprotective effect in a rat model of focal ischemia and that Isradipine produces a dose-dependent sparing of dopamine neuronal fibers and cell bodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isradipine is a L-type calcium channel blocker, Isradipine has anti-atherosclerotic effects in animals and improves endothelium-mediated nitric oxide (NO)-dependent vasodilation in vitro. Additional studies indicate that Isradipine may provide a neuroprotective effect in a rat model of focal ischemia and that Isradipine produces a dose-dependent sparing of dopamine neuronal fibers and cell bodies.

Solubility
Soluble in DMSO

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75695-93-1
Molecular Formula C19H21N3O5
Molecular Weight (g/mol) 371.39
MDL Number MFCD00153820
InChI Key HMJIYCCIJYRONP-UHFFFAOYNA-N
Synonym isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine
PubChem CID 3784
IUPAC Name 3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C

Specifications

Color Yellow
Quantity 50 mg
Merck Index 14,5240
Solubility Information Soluble in DMSO
Formula Weight 371.39
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Isradipine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.