missing translation for 'onlineSavingsMsg'
Learn More

Isotachysterol3, 90%, TRC

Product Code. 18051930
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18051930 25 mg 25mg
18042940 5 mg 5mg
18023040 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18051930 Supplier Toronto Research Chemicals Supplier No. TRCI90246025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22350-43-2
Molecular Formula C27 H44 O
Molecular Weight (g/mol) 384.64
Synonym 9,10-Secocholesta-5(10),6,8(14)-trien-3-ol, (3β,6E)- (9CI), 9,10-Secocholesta-5(10),6,8(14)-trien-3β-ol, (E)- (8CI), Isotachysterol3 (6CI), Toxisterol3 D3, (6E)-9,10-Secocholesta-5(10),6,8(14)-trien-3β-ol, Isotachysterol3
IUPAC Name (1S)-3-[(E)-2-[(1R,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-1,2,3,5,6,7-hexahydroinden-4-yl]ethenyl]-4-methylcyclohex-3-en-1-ol
SMILES CC(C)CCC[C@@H](C)[C@H]1CCC2=C(CCC[C@]12C)\C=C\C3=C(C)CC[C@H](O)C3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-25,28H,6-11,14-18H2,1-5H3/b13-12+/t21-,24+,25-,27-/m1/s1
Formula Weight 384.3392
Chemical Name or Material Isotachysterol 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.