CAS RN 22350-43-2
CAS RN 22350-43-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isotachysterol3, 90%, TRC
CAS: 22350-43-2 Molecular Formula: C27 H44 O Molecular Weight (g/mol): 384.64 Synonym: 9,10-Secocholesta-5(10),6,8(14)-trien-3-ol, (3β,6E)- (9CI),9,10-Secocholesta-5(10),6,8(14)-trien-3β-ol, (E)- (8CI),Isotachysterol3 (6CI),Toxisterol3 D3,(6E)-9,10-Secocholesta-5(10),6,8(14)-trien-3β-ol,Isotachysterol3 IUPAC Name: (1S)-3-[(E)-2-[(1R,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-1,2,3,5,6,7-hexahydroinden-4-yl]ethenyl]-4-methylcyclohex-3-en-1-ol SMILES: CC(C)CCC[C@@H](C)[C@H]1CCC2=C(CCC[C@]12C)\C=C\C3=C(C)CC[C@H](O)C3