missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide mononitrate, 98+%, Thermo Scientific™

A coronary vasodilator that does not require hepatic biotransformation

£223.00 - £530.00

Chemical Identifiers

CAS 16051-77-7
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.139
MDL Number MFCD00143462
InChI Key YWXYYJSYQOXTPL-SLPGGIOYSA-N
Synonym isosorbide mononitrate, isosorbide 5-mononitrate, monosorbitrate, imdur, elantan, monoket, mononit, corangin, ismn, isosorbide 5-nitrate
PubChem CID 27661
ChEBI CHEBI:6062
IUPAC Name [(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
SMILES C1C(C2C(O1)C(CO2)O[N+](=O)[O-])O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15405859
SDS View Documents
Thermo Scientific Alfa Aesar
J63674.09
10g
£223.00
10g
View alternative products
Save Up to 
»
Estimated Shipment: 12-02-2027
Add to basket
 
 Restricted Item
15415859
SDS View Documents
Thermo Scientific Alfa Aesar
J63674.14
25g
£530.00
25g
View alternative products
Save Up to 
»
Estimated Shipment: 02-10-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

16051-77-7
191.139
YWXYYJSYQOXTPL-SLPGGIOYSA-N
27661
[(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
C6H9NO6
MFCD00143462
isosorbide mononitrate, isosorbide 5-mononitrate, monosorbitrate, imdur, elantan, monoket, mononit, corangin, ismn, isosorbide 5-nitrate
CHEBI:6062
C1C(C2C(O1)C(CO2)O[N+](=O)[O-])O

Specifications

16051-77-7
MFCD00143462
14,5225
YWXYYJSYQOXTPL-SLPGGIOYSA-N
[(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
27661
191.14
Isosorbide mononitrate
C6H9NO6
UN3251
isosorbide mononitrate, isosorbide 5-mononitrate, monosorbitrate, imdur, elantan, monoket, mononit, corangin, ismn, isosorbide 5-nitrate
C1C(C2C(O1)C(CO2)O[N+](=O)[O-])O
191.139
CHEBI:6062
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: ISOSORBIDE-5-MONONITRATE

missing translation for 'einecsNumber' : 240-197-2

missing translation for 'rtecsNumber' : LZ4386500

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.