missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide mononitrate, 98+%, Thermo Scientific™

A coronary vasodilator that does not require hepatic biotransformation

Brand:  Thermo Scientific Alfa Aesar J63674.09

Quantity 10g

 View more versions of this product

Product Code. 15405859

  • £223.00 / 10g
 Restricted Item
Estimated Shipment: 12-02-2027
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

16051-77-7
191.139
YWXYYJSYQOXTPL-SLPGGIOYSA-N
27661
[(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
C6H9NO6
MFCD00143462
isosorbide mononitrate, isosorbide 5-mononitrate, monosorbitrate, imdur, elantan, monoket, mononit, corangin, ismn, isosorbide 5-nitrate
CHEBI:6062
C1C(C2C(O1)C(CO2)O[N+](=O)[O-])O

Specifications

16051-77-7
MFCD00143462
UN3251
isosorbide mononitrate, isosorbide 5-mononitrate, monosorbitrate, imdur, elantan, monoket, mononit, corangin, ismn, isosorbide 5-nitrate
C1C(C2C(O1)C(CO2)O[N+](=O)[O-])O
191.139
CHEBI:6062
≥98%
C6H9NO6
10g
14,5225
YWXYYJSYQOXTPL-SLPGGIOYSA-N
[(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
27661
191.14
Isosorbide mononitrate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.