missing translation for 'onlineSavingsMsg'
Learn More

Isopropylmagnesium chloride - LiCl complex, 1M in MeTHF

Product Code. 15459205
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459205 500 mL 500mL
15449205 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459205 Supplier Thermo Scientific Alfa Aesar Supplier No. H51156.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 807329-97-1 | C3H7Cl2LiMg | 145.23 g/mol

Isopropylmagnesium chloride is used in the regioselective functionalization of trisubstituted pyridines, electrophilic amination for preparation of amines and functionalization of cyclopentene derivatives. It is involved in the preparation of 2,3-functionalized furans, benzofurans, and thiophenes. Further, it is used in benzylic Grignards via sulfur-magnesium exchange.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylmagnesium chloride is used in the regioselective functionalization of trisubstituted pyridines, electrophilic amination for preparation of amines and functionalization of cyclopentene derivatives. It is involved in the preparation of 2,3-functionalized furans, benzofurans, and thiophenes. Further, it is used in benzylic Grignards via sulfur-magnesium exchange.

Solubility
Miscible with ethers and amines.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 807329-97-1
Molecular Formula C3H7Cl2LiMg
Molecular Weight (g/mol) 145.23
MDL Number MFCD07784514
InChI Key CWTUREABAILGIK-UHFFFAOYSA-L
Synonym turbo grignard, iprmgcl licl, i-prmgcl licl, i-prmgcl.licl, isopropylmagnesiumchloride licl, isopropylmagnesium chloride licl, isopropyl magnesium chloride licl, cwtureabailgik-uhfffaoysa-l, isopropylmagnesium lithium chloride, isopropyl magnesium chloride li-cl
IUPAC Name lithium(1+) chloro(propan-2-yl)magnesium chloride
SMILES [Li+].[Cl-].CC(C)[Mg]Cl

Specifications

Density 0.951
Flash Point −17°C (1.4°F)
Linear Formula (CH3)2CHMgCl·LiCl
Quantity 500 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Miscible with ethers and amines.
Formula Weight 145.24
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material Isopropylmagnesium chloride - LiCl complex

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.