CAS RN 807329-97-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isopropylmagnesium chloride - LiCl complex, 1M in MeTHF
CAS: 807329-97-1 Molecular Formula: C3H7Cl2LiMg Molecular Weight (g/mol): 145.23 MDL Number: MFCD07784514 InChI Key: CWTUREABAILGIK-UHFFFAOYSA-L Synonym: turbo grignard,iprmgcl licl,i-prmgcl licl,i-prmgcl.licl,isopropylmagnesiumchloride licl,isopropylmagnesium chloride licl,isopropyl magnesium chloride licl,cwtureabailgik-uhfffaoysa-l,isopropylmagnesium lithium chloride,isopropyl magnesium chloride li-cl IUPAC Name: lithium(1+) chloro(propan-2-yl)magnesium chloride SMILES: [Li+].[Cl-].CC(C)[Mg]Cl