missing translation for 'onlineSavingsMsg'
Learn More

Isopropylmagnesium chloride - LiCl complex, 1M in MeTHF

Product Code. 15449205
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449205 100 mL 100mL
15459205 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449205 Supplier Thermo Scientific Alfa Aesar Supplier No. H51156.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 807329-97-1 | C3H7Cl2LiMg | 145.23 g/mol

Isopropylmagnesium chloride is used in the regioselective functionalization of trisubstituted pyridines, electrophilic amination for preparation of amines and functionalization of cyclopentene derivatives. It is involved in the preparation of 2,3-functionalized furans, benzofurans, and thiophenes. Further, it is used in benzylic Grignards via sulfur-magnesium exchange.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylmagnesium chloride is used in the regioselective functionalization of trisubstituted pyridines, electrophilic amination for preparation of amines and functionalization of cyclopentene derivatives. It is involved in the preparation of 2,3-functionalized furans, benzofurans, and thiophenes. Further, it is used in benzylic Grignards via sulfur-magnesium exchange.

Solubility
Miscible with ethers and amines.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 807329-97-1
Molecular Formula C3H7Cl2LiMg
Molecular Weight (g/mol) 145.23
MDL Number MFCD07784514
InChI Key CWTUREABAILGIK-UHFFFAOYSA-L
Synonym turbo grignard, iprmgcl licl, i-prmgcl licl, i-prmgcl.licl, isopropylmagnesiumchloride licl, isopropylmagnesium chloride licl, isopropyl magnesium chloride licl, cwtureabailgik-uhfffaoysa-l, isopropylmagnesium lithium chloride, isopropyl magnesium chloride li-cl
IUPAC Name lithium(1+) chloro(propan-2-yl)magnesium chloride
SMILES [Li+].[Cl-].CC(C)[Mg]Cl

Specifications

Density 0.951
Flash Point −17°C (1.4°F)
Linear Formula (CH3)2CHMgCl·LiCl
Quantity 100 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Miscible with ethers and amines.
Formula Weight 145.24
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material Isopropylmagnesium chloride - LiCl complex
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Isopropylmagnesium chloride - LiCl complex, 1M in MeTHF
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which, in contact with water, emits flammable gases Category 1
Faresætning
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
Supplerende oplysninger
  • MIXTURE LIST-Contain: 2-Methyltetrahydrofuran, Isopropylmagnesium chloride

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.