missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 3-acetylpyridine-2-carboxylate, 97%, Thermo Scientific™

Product Code. 11363157
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363157 25 g 25g
11353157 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363157 Supplier Thermo Scientific Alfa Aesar Supplier No. L13583.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195812-68-1
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD01314548
InChI Key KTGMAKWQZOTIDD-UHFFFAOYSA-N
Synonym isopropyl 3-acetylpyridine-2-carboxylate, isopropyl 3-acetylpicolinate, methylethyl 3-acetylpyridine-2-carboxylate, acmc-20akow, isopropyl3-acetylpyridine-2-carboxylate, 3-acetylpyridine-2-carboxylic acid isopropyl ester, 2-pyridinecarboxylicacid,3-acetyl-,1-methylethyl ester
PubChem CID 3757842
IUPAC Name propan-2-yl 3-acetylpyridine-2-carboxylate
SMILES CC(C)OC(=O)C1=C(C=CC=N1)C(C)=O

Specifications

Melting Point 38°C to 42°C
Boiling Point 140°C to 142°C (1 mmHg)
Quantity 25 g
Beilstein 7760203
Formula Weight 207.23
Percent Purity 97%
Chemical Name or Material Isopropyl 3-acetylpyridine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.