accessibility menu, dialog, popup

UserName

H3CCH3OONOCH3

CAS RN 195812-68-1

IUPAC Name: propan-2-yl 3-acetylpyridine-2-carboxylate
Synonyms: isopropyl 3-acetylpyridine-2-carboxylate
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: KTGMAKWQZOTIDD-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C1=C(C=CC=N1)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results