missing translation for 'onlineSavingsMsg'
Learn More

Isopropenyl chloroformate, 96%, stabilized

Product Code. 10308870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10308870 5 g Glass bottle 5g
10539253 1 g Glass bottle 1g
10534474 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10308870 Supplier Thermo Scientific Acros Supplier No. 275120050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 57933-83-2 | C4H5ClO2 | 120.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57933-83-2
MDL Number MFCD00037083
InChI Key AMMGCGVWJMRTQI-UHFFFAOYSA-N
Synonym isopropenyl chloroformate, carbonochloridic acid, 1-methylethenyl ester, isopropenylchloroformate, isopropenylchloro-formate, isopropenyl carbonochloridate, isopropenyl chloridocarbonate, prop-1-en-2-yl chloroformate, chloroformic acid isopropenyl ester, chloroformic acid,isopropenyl ester, chloro prop-1-en-2-yloxy methanone
PubChem CID 93846
IUPAC Name prop-1-en-2-yl carbonochloridate
SMILES CC(=C)OC(=O)Cl

Specifications

Color Colorless
Density 1.1030g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C4H5ClO2
Linear Formula CH2=C(CH3)CO2Cl
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
Quantity 5 g
Stabilizer 50 to 100ppm BHT
Molecular Weight (g/mol) 120.532
Formula Weight 120.54
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Isopropenyl chloroformate, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isopropenyl chloroformate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.