accessibility menu, dialog, popup

UserName

CH3H2COClO

CAS RN 57933-83-2

IUPAC Name: prop-1-en-2-yl carbonochloridate
Synonyms: chloroformic acid,isopropenyl ester
Molecular Weight (g/mol): 120.53
Molecular Formula: C4H5ClO2
InChi Key: AMMGCGVWJMRTQI-UHFFFAOYSA-N
SMILES: CC(=C)OC(Cl)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results