missing translation for 'onlineSavingsMsg'
Learn More

Isometheptene-d3 Maleate, TRC

Product Code. 30396941
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396941 50 mg
30389668 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30396941 Supplier Toronto Research Chemicals Supplier No. TRCI82132750MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1795136-87-6
Molecular Formula C13H20D3NO4
Molecular Weight (g/mol) 260.34
Synonym N-(Methyl-d3)-6-methyl-5-hepten-2-amine (2Z)-2-Butenedioate, N-(Methyl-d3)-6-methyl-5-hepten-2-amine (Z)-2-Butenedioate
IUPAC Name (Z)-but-2-enedioic acid;6-methyl-N-(trideuteriomethyl)hept-5-en-2-amine
SMILES [2H]C([2H])([2H])NC(C)CCC=C(C)C.OC(=O)\C=C/C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H19N.C4H4O4/c1-8(2)6-5-7-9(3)10-4;5-3(6)1-2-4(7)8/h6,9-10H,5,7H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/i4D3;
Formula Weight 260.1815
Chemical Name or Material Isometheptene-d3 Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.