CAS RN 1795136-87-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isometheptene-d3 Maleate, TRC
CAS: 1795136-87-6 Molecular Formula: C13H20D3NO4 Molecular Weight (g/mol): 260.34 Synonym: N-(Methyl-d3)-6-methyl-5-hepten-2-amine (2Z)-2-Butenedioate,N-(Methyl-d3)-6-methyl-5-hepten-2-amine (Z)-2-Butenedioate IUPAC Name: (Z)-but-2-enedioic acid;6-methyl-N-(trideuteriomethyl)hept-5-en-2-amine SMILES: [2H]C([2H])([2H])NC(C)CCC=C(C)C.OC(=O)\C=C/C(=O)O