missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Isolator Wrap Sterile Contact Sabouraud Dextrose 5.6 Agar Plates with Lecithin & Polysorbate 80 - Room Temperature
GREENER_CHOICE

Product Code. 17782608
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17782608 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17782608 Supplier Thermo Scientific™ Supplier No. R111825RT
  • ONLINE EXCLUSIVE PRICE
    £97.85
    £92.68 / Pack of 10
    Save £5.17
    5% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate with Lecithin & Polysorbate 80 is used for the cultivation of yeasts and fungi.

  • Settling pink plates (single bag and double bag) are VHP resistant manufactured in a clean room environment. This provides the highest level of sterility assurance with no compromise of growth characteristics.
  • Each lot is tested to ensure consistent, reproducible results.
  • Storage Condition: 2 to 25°C.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 20°C to 25°C, Room Temperature
Media Type Sabouraud Dextrose Agar
Product Type Contact Plate
For Use With (Equipment) RABS, Isolators and Cleanrooms
Quantity 10/Pk.
Diameter (Metric) 85 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.