missing translation for 'onlineSavingsMsg'
Learn More

Isoguanosine Hydrate, TRC

Product Code. 30391937
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391937 10 mg 10mg
30392642 50 mg -
30403321 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391937 Supplier Toronto Research Chemicals Supplier No. TRCI81950010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 359436-55-8
Molecular Formula C10 H13 N5 O5 . H2 O
Molecular Weight (g/mol) 301.26
IUPAC Name 6-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-2-one;hydrate
SMILES O.NC1=NC(=O)Nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H13N5O5.H2O/c11-7-4-8(14-10(19)13-7)15(2-12-4)9-6(18)5(17)3(1-16)20-9;/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19);1H2/t3-,5-,6-,9-;/m1./s1
Formula Weight 301.1022
Chemical Name or Material Isoguanosine Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.