CAS RN 359436-55-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isoguanosine Hydrate, TRC
CAS: 359436-55-8 Molecular Formula: C10 H13 N5 O5 . H2 O Molecular Weight (g/mol): 301.26 IUPAC Name: 6-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-2-one;hydrate SMILES: O.NC1=NC(=O)Nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O