missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Iso-Sensitest Agar

Product Code. 12902638
Change view
Click to view available options
Description:
Iso-Sensitest Agar
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity unitSize
12902638 Iso-Sensitest Agar 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12902638 Supplier Thermo Scientific™ Supplier No. PO0779A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform effortless testing using a semi-defined medium specifically designed for antimicrobial susceptibility testing (AST).

Perform effortless testing using Thermo Scientific™ Iso-Sensitest Agar, a semi-defined medium specifically designed for antimicrobial susceptibility testing (AST).

Iso-Sensitest Agar has stability in mineral content and the ability to produce optimum antimicrobial zones of inhibition. The amino-nitrogen base of acid-hydrolysed casein and special peptones has been supplemented with defined growth factors. Careful preparation of the nutrients ensures that trimethoprim and sulphonamide antagonists are at very low levels.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance According to BSAC
Color Straw, clear
Content And Storage 2°C to 10°C
Description Iso-Sensitest Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 26g ± 0.5g
Packaging Type 4mm depth, wrapped in single cellulose-based film wrap
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.