missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ISO-Sensitest Agar
Description
This specialized agar medium is designed to support the accurate determination of microbial resistance, ensuring reliable results for your microbiological testing needs.
Specifications
Specifications
| Color | Transparent |
| Content And Storage | 2°C to 12°C |
| Format | 90 mm |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Volume | 17 mL |
| Packaging Type | 1 x 10 Plates |
| Quantity | 10 Plates |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?