missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Islet-1 Antibody (4F6), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Islet-1 |
|---|---|
| Clone | 4F6 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30027503
|
Novus Biologicals
NBP3-26191-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30034620
|
Novus Biologicals
NBP3-26191-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Islet-1 Monoclonal antibody specifically detects Islet-1 in Human samples. It is validated for Western Blot, ELISASpecifications
| Islet-1 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| insulin gene enhancer protein ISL-1, ISL LIM homeobox 1, Isl-1, ISL1 transcription factor, LIM/homeodomain, ISL1 transcription factor, LIM/homeodomain, (islet-1), Islet-1, ISLET1 | |
| A synthesized peptide derived from Human Islet-1 [UniProt P61371] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 4F6 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3670 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title