missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Islet-1 Antibody (4F6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26191-100ul
This item is not returnable.
View return policy
Description
Islet-1 Monoclonal antibody specifically detects Islet-1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Islet-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4F6 | |
| Western Blot 1:500-1:5000, ELISA | |
| insulin gene enhancer protein ISL-1, ISL LIM homeobox 1, Isl-1, ISL1 transcription factor, LIM/homeodomain, ISL1 transcription factor, LIM/homeodomain, (islet-1), Islet-1, ISLET1 | |
| A synthesized peptide derived from Human Islet-1 [UniProt P61371] | |
| 100 μL | |
| Diabetes Research, Lipid and Metabolism, Stem Cell Markers | |
| 3670 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction