missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRX3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£378.00
Specifications
| Antigen | IRX3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
IRX3 Polyclonal specifically detects IRX3 in Mouse samples. It is validated for Western Blot.Specifications
| IRX3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Homeodomain protein IRXB1, iroquois homeobox 3, Iroquois homeobox protein 3, iroquois-class homeodomain protein IRX-1, iroquois-class homeodomain protein IRX-3, IRX-1, IRXB1FLJ99187 | |
| IRX3 | |
| IgG | |
| Protein A purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| NP_032419 | |
| 79191 | |
| Synthetic peptide directed towards the N terminal of mouse IRX3. Peptide sequence LGYQYIRPLYPPERPGAAGGGGGGSSAGGRSGPGAGASELAASGSLSNVL. | |
| Primary | |
| 56 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title