missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRX3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80217
This item is not returnable.
View return policy
Description
IRX3 Polyclonal specifically detects IRX3 in Mouse samples. It is validated for Western Blot.
Specifications
| IRX3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Homeodomain protein IRXB1, iroquois homeobox 3, Iroquois homeobox protein 3, iroquois-class homeodomain protein IRX-1, iroquois-class homeodomain protein IRX-3, IRX-1, IRXB1FLJ99187 | |
| Rabbit | |
| 56 kDa | |
| 100 μL | |
| Primary | |
| Rat: 86%; Porcine: 79%. | |
| Mouse, Rat, Pig, Bovine, Canine | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_032419 | |
| IRX3 | |
| Synthetic peptide directed towards the N terminal of mouse IRX3. Peptide sequence LGYQYIRPLYPPERPGAAGGGGGGSSAGGRSGPGAGASELAASGSLSNVL. | |
| Protein A purified | |
| RUO | |
| 79191 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction