missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£158.00 - £348.00
Specifications
| Antigen | IRS1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18319475
|
Novus Biologicals
NBP3-16898-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18395125
|
Novus Biologicals
NBP3-16898-100UL |
100 μg |
£348.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRS1 Polyclonal antibody specifically detects IRS1 in Human samples. It is validated for Western BlotSpecifications
| IRS1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| HIRS-1, insulin receptor substrate 1, IRS-1 | |
| A synthetic phosphorylated peptide around S636 & S639 of human IRS1 (NP_005535.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Neuroscience, Phospho Specific, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 3667 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title