Learn More
Chemsplash Irradiated Zip Labcoat PB(6B) Thumb Loops, Elastic Cuff
GREENER_CHOICE
Product Code. p-200105915
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
missing translation for 'unitSize'
Pack of 50
Product Code. 30058345
missing translation for 'mfr': Chemsplash 2637.400.004
Only null left
Add to basket
Add to basket
Supplied gamma Sterile Irradiated and double bagged. This lab coat is a 2/3 length coat with a collar and zip closure and elasticated wrists with thumb loops.
Chemsplash Lab Coat
  • Chemsplash 55GSM Type PB 6B Fabric
  • GAMMA Irradiated and Double Bagged
  • Zip Fasten Front with Adhesive Flap Cover
  • Elasticated Cuffs with Thumb Loops
  • Fold Over Collar
  • Silicone and Latex Free, PFAS Free
  • Anti-Static Material
  • Non-Linting
  • Conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9
Description: The Chemsplash Lab Coat is supplied GAMMA Sterile Irradiated and double bagged. Made from Chemsplash fabric, it is soft and microporous, offering strength and resistance to chemical splash and dust. This lab coat features a 2/3 length design with a collar and zip closure, as well as elasticated wrists with thumb loops for added convenience. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9. Chemsplash accessories protect the wearer from the environment or the environment from the wearer, designed for maximum comfort and wearability.
Specifications
Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Waist Style Loose
Ankle Style Open
Cuff Style Elastic, Thumb Loop
Neck Style Collared
Hood Type Standard Collar
Closure Type Ties
Seam Type Stitched
Sterility Sterile
Resistance Liquid
Latex Free Yes
No. of Pockets 1
Cleanroom Class Class 100-100,000 (ISO 5-8)