missing translation for 'onlineSavingsMsg'
Learn More

Iron yttrium oxide, 99.9% (REO)

Product Code. 11338288
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338288 25 g 25g
11348288 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338288 Supplier Thermo Scientific Alfa Aesar Supplier No. 039488.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12063-56-8 | Fe5O12Y3 | 737.931 g/mol

Iron yttrium oxide is mainly used in superconductors. It is also used in filters, transmitters, transducers, solid state lasers in faraday rotators and in non-linear optics. It finds application in polymers, textiles, fuel cell layers, composites and solar energy materials, bio nano materials and bio sensors. Further, it is used in the preparation of fuel cells and solar energy materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12063-56-8
Molecular Formula Fe5O12Y3
Molecular Weight (g/mol) 737.931
MDL Number MFCD00210618
InChI Key UVXIKKWNYGPENJ-UHFFFAOYSA-N
Synonym iron yttrium oxide, yttrium iron garnet, yttrium ferrite garnet, bis ferric oxide ; oxo oxoyttrio oxy iron; oxo oxoyttrio oxy yttrium, yttrium iron oxide, nanopowder, <100 nm particle size bet trace metals basis
PubChem CID 16217670
IUPAC Name oxo(oxoferriooxy)iron;oxo(oxoferriooxy)yttrium;oxo(oxoyttriooxy)yttrium
SMILES O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Y]=O.O=[Y]O[Y]=O

Specifications

Color Green
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 737.94
Odor Odorless
Appearance Green powder
Chemical Name or Material Iron yttrium oxide
compliance-icons
Product Identifier
  • Iron yttrium oxide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.