CAS RN 12063-56-8
IUPAC Name:
iron(3+) yttrium(3+) trioxidandiide
Synonyms:
iron(3+) yttrium(3+) trioxidandiide
Molecular Weight (g/mol):
192.75
Molecular Formula:
FeO3Y
InChi Key:
SUFJITMMSUGKIE-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[Fe+3].[Y+3]
1
–
1
of
1
results
1
Iron yttrium oxide, 99.9% (REO)
CAS: 12063-56-8 Molecular Formula: Fe5O12Y3 Molecular Weight (g/mol): 737.931 MDL Number: MFCD00210618 InChI Key: UVXIKKWNYGPENJ-UHFFFAOYSA-N Synonym: iron yttrium oxide,yttrium iron garnet,yttrium ferrite garnet,bis ferric oxide ; oxo oxoyttrio oxy iron; oxo oxoyttrio oxy yttrium,yttrium iron oxide, nanopowder, <100 nm particle size bet trace metals basis PubChem CID: 16217670 IUPAC Name: oxo(oxoferriooxy)iron;oxo(oxoferriooxy)yttrium;oxo(oxoyttriooxy)yttrium SMILES: O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Y]=O.O=[Y]O[Y]=O