missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron nickel oxide, tech., Thermo Scientific Chemicals
30.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
Iron nickel oxide is used as an adsorbent to remove hexavalent chromium ions present in the waste water by co-sedimentation method. It is also employed as an additive in membrane (i.e. polyvinylchloride/styrene-butadiene-rubber blend heterogeneous cation exchange membrane) fabrication, thereby increasing the permeability for bivalent ions compared to the monovalent type.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown |
Physical Form | Powder |
Quantity | 10 g |
Formula Weight | 234.39 |
Grade | Technical |
Odor | Odorless |
Appearance | Dark-brown powder |
Density | 5.368 |
Chemical Name or Material | Iron nickel oxide |
Safety and Handling
Product Identifier
- Iron nickel oxide
- Danger
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P363-Wash contaminated clothing before reuse.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'